L(+)-Glutamic acid 98%

Supplier: Ambeed
Warning

Synonyms: (S)-(+)-2-Aminopentanedioic acid, L(+)-1-Aminopropane-1,3-dicarboxylic acid, (S)-(+)-α-Glutamic acid, H-Glu-OH, L-Glutamic acid, L(+)-2-Aminopentanedioic acid, (S)-(+)-Glutamic acid, (S)-(+)-1-Aminopropane-1,3-dicarboxylic acid, L(+)-α-Glutamic acid

A211917-1000G A211917-100G A211917-25G A211917-500G
77164-966EA 17 USD
77164-966 77164-968 77164-970 77164-972
L(+)-Glutamic acid 98%
L(+)-Glutamic acid

Caution: for Research Use Only

Formula: HO₂CCH₂CH₂CH(NH₂)CO₂H
MW: 147.13 g/mol
Boiling Pt: 325
Melting Pt: 160
MDL Number: MFCD00002634
CAS Number: 56-86-0
Merck Index: 13,04482

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR