Ethyl-trans-4-hydroxycyclohexanecarboxylate 98%, Ambeed.Inc

Supplier: Ambeed
Warning

A200448-100G A200448-1G A200448-5G A200448-25G A200448-10G
77164-572EA 155 USD
77164-572 77164-574 77164-576 77567-892 77683-559
Ethyl-trans-4-hydroxycyclohexanecarboxylate 98%, Ambeed.Inc
Ethyl-trans-4-hydroxycyclohexanecarboxylate

Caution: For research use only.

Formula: C₉H₁₆O₃
MDL Number: MFCD16295187
CAS Number: 3618-04-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR