Pirfenidone 98%

Supplier: Ambeed
Warning

Synonyms: 5-Methyl-1-phenylpyridin-2(1H)-one, 5-Methyl-1-phenyl-2-pyridone

A196927-100G A196927-100MG A196927-10G A196927-1G A196927-25G A196927-5G
77164-448EA 414.99 USD
77164-448 77164-450 77164-452 77164-454 77164-456 77164-458 77164-460
Pirfenidone 98%
Pirfenidone

Caution: for Research Use Only

Formula: C₁₂H₁₁NO
MW: 185.23 g/mol
MDL Number: MFCD00866047
CAS Number: 53179-13-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR