Pirfenidone 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Methyl-1-phenylpyridin-2(1H)-one, 5-Methyl-1-phenyl-2-pyridone
A196927-100G
A196927-100MG
A196927-10G
A196927-1G
A196927-25G
A196927-5G
77164-448EA
414.99
USD
77164-448
77164-450
77164-452
77164-454
77164-456
77164-458
77164-460
Pirfenidone 98%
Pirfenidone
Caution: for Research Use Only
Formula:
C₁₂H₁₁NO MW: 185.23 g/mol |
MDL Number:
MFCD00866047 CAS Number: 53179-13-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...