N-Methyl-D-aspartic acid 98%

Supplier: Ambeed
Warning

A188283-100MG A188283-1MG A188283-50MG
77164-142EA 38 USD
77164-142 77164-144 77164-146
N-Methyl-D-aspartic acid 98%
N-Methyl-D-aspartic acid

Caution: for Research Use Only

Formula: C₅H₉NO₄
MDL Number: MFCD00004226
CAS Number: 6384-92-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR