5-Bromo-2-fluoroisonicotinic acid 95%
Supplier: Ambeed
|
Warning
|
A186957-1G
A186957-250MG
A186957-5G
77164-094EA
43
USD
77164-094
77164-096
77164-098
5-Bromo-2-fluoroisonicotinic acid 95%
5-Bromo-2-fluoroisonicotinic acid
Caution: for Research Use Only
Formula:
C₆H₃BrFNO₂ MW: 220 g/mol |
MDL Number:
MFCD13185803 CAS Number: 1214377-40-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...