Empagliflozin 98%

Supplier: Ambeed
Warning

Synonyms: (2S,3R,4R,5S,6R)-2-(4-Chloro-3-(4-(((S)-tetrahydrofuran-3-yl)oxy)benzyl)phenyl)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol, Jardiance

A112500-10MG A112500-1G A112500-250MG A112500-50MG A112500-5G A112500-10G A112500-100MG
77160-806EA 5 USD
77160-806 77160-808 77160-810 77160-812 77160-814 77570-690 77891-810
Empagliflozin 98%
Empagliflozin

Caution: for Research Use Only

Formula: C₂₃H₂₇ClO₇
MDL Number: MFCD22566222
CAS Number: 864070-44-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR