2-Hydroxypyridine 98%

Supplier: Ambeed
Danger

Synonyms: Pyridin-2-one, Pyridin-2(1H)-one, α-Hydroxypyridine, 2-Pyridone, α-Pyridone

A913349-10G A913349-25G A913349-100G
77159-856EA 13 USD
77159-856 77159-858 77159-860 77614-634
2-Hydroxypyridine 98%
2-Hydroxypyridine

Caution: for Research Use Only

Formula: C₅H₅NO
MDL Number: MFCD00006268
CAS Number: 142-08-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR