Methyl-4-fluoro-5-methoxy-2-nitrobenzoate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 4-fluoro-5-methoxy-2-nitrobenzoate
A814077-1G
A814077-250MG
A814077-5G
77159-302EA
20
USD
77159-302
77159-304
77159-306
Methyl-4-fluoro-5-methoxy-2-nitrobenzoate 98%
Methyl-4-fluoro-5-methoxy-2-nitrobenzoate
Caution: for Research Use Only
Formula:
C₉H₈FNO₅ |
MDL Number:
MFCD20921594 CAS Number: 159768-50-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...