IQ-1 98+%

Supplier: Ambeed
Warning

Synonyms: 2-((4-Acetylphenyl)diazenyl)-2-(3,3-dimethyl-3,4-dihydroisoquinolin-1(2H)-ylidene)acetamide, Wnt Pathway Activator III, IQ 1, 2-(4-Acetylphenylazo)-2-(3,3-dimethyl-3,4-dihydro-2H-isoquinolin-1-ylidene)-acetamide

A757329-10MG A757329-1MG A757329-25MG A757329-50MG A757329-5MG
77158-928EA 71 USD
77158-928 77158-930 77158-932 77158-934 77158-936
IQ-1 98+%
IQ-1

Caution: for Research Use Only

Formula: C₂₁H₂₂N₄O₂
MW: 362.43 g/mol
MDL Number: MFCD00354369
CAS Number: 331001-62-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR