Benzophenone 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
Diphenylmethanone, Diphenyl ketone
A704460-25G
A704460-500G
A704460-1000G
A704460-100G
77158-472EA
5.15
USD
77158-472
77158-474
77552-138
77573-286
77573-288
Benzophenone 98%
Benzophenone
Caution: for Research Use Only
Formula:
C₁₃H₁₀O |
MDL Number:
MFCD00003076 CAS Number: 119-61-9 UN: 3077 ADR: 9,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...