PF-04217903 98+%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-(4-(1-(Quinolin-6-ylmethyl)-1H-[1,2,3]triazolo[4,5-b]pyrazin-6-yl)-1H-pyrazol-1-yl)ethan-1-ol
A670353-10MG
A670353-1MG
A670353-50MG
A670353-5MG
77158-150EA
64
USD
77158-150
77158-152
77158-154
77158-156
PF-04217903 98+%
PF-04217903
Caution: for Research Use Only
Formula:
C₁₉H₁₆N₈O |
MDL Number:
MFCD12407410 CAS Number: 956905-27-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...