PF-04217903 98+%

Supplier: Ambeed
Warning

Synonyms: 2-(4-(1-(Quinolin-6-ylmethyl)-1H-[1,2,3]triazolo[4,5-b]pyrazin-6-yl)-1H-pyrazol-1-yl)ethan-1-ol

A670353-10MG A670353-1MG A670353-50MG A670353-5MG
77158-150EA 64 USD
77158-150 77158-152 77158-154 77158-156
PF-04217903 98+%
PF-04217903

Caution: for Research Use Only

Formula: C₁₉H₁₆N₈O
MDL Number: MFCD12407410
CAS Number: 956905-27-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR