1-Bromo-3,4,5-trifluorobenzene 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
5-Bromo-1,2,3-trifluorobenzene
A527209-10G
A527209-5G
A527209-25G
A527209-100G
77156-968EA
6
USD
77156-968
77156-970
77551-590
77613-656
1-Bromo-3,4,5-trifluorobenzene 98%
1-Bromo-3,4,5-trifluorobenzene
Caution: for Research Use Only
Formula:
C₆H₂BrF₃ MW: 210.98 Boiling Pt: 140 Melting Pt: -20 Density: 1.767 |
MDL Number:
MFCD00070738 CAS Number: 138526-69-9 UN: 1993 ADR: 3 Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...