Wedelolactone 98%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: 1,8,9-Trihydroxy-3-methoxy-6H-benzofuro[3,2-c]chromen-6-one, 7-Methoxy-5,11,12-trihydroxy-coumestan

A523974-10MG A523974-1MG A523974-5MG A523974-1G A523974-100MG A523974-250MG A523974-50MG
77156-928EA 39 USD
77156-928 77156-930 77156-932 77684-356 77893-070 77893-071 77893-072
Wedelolactone 98%, Ambeed.Inc
Wedelolactone

Caution: For research use only.

Formula: C₁₆H₁₀O₇
MDL Number: MFCD07778564
CAS Number: 524-12-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR