(S)-(-)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl 98%

Supplier: Ambeed
Warning

Synonyms: (S)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl, (S)-(-)-TolBINAP

A491448-10G A491448-1G A491448-250MG A491448-5G A491448-25G
77156-572EA 281 USD
77156-572 77156-574 77156-576 77156-578 77901-816
(S)-(-)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl 98%
(S)-(-)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl

Caution: for Research Use Only

Formula: C₄₈H₄₀P₂
MDL Number: MFCD01311709
CAS Number: 100165-88-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR