1-Butyl-3-methylimidazolium methanesulfonate 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
1-n-Butyl-3-methylimidazolium methanesulfonate, 1-n-Butyl-3-methylimidazolium methanesulphonate
A442178-1G
A442178-25G
A442178-5G
77156-068EA
14
USD
77156-068
77156-070
77156-072
1-Butyl-3-methylimidazolium methanesulfonate 98%
1-Butyl-3-methylimidazolium methanesulphonate
Caution: for Research Use Only
Formula:
C₉H₁₈N₂O₃S |
MDL Number:
MFCD06798173 CAS Number: 342789-81-5 UN: 2923 ADR: 8,6.1,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...