1-Butyl-3-methylimidazolium methanesulfonate 98%

Supplier: Ambeed
Danger

Synonyms: 1-n-Butyl-3-methylimidazolium methanesulfonate, 1-n-Butyl-3-methylimidazolium methanesulphonate

A442178-1G A442178-25G A442178-5G
77156-068EA 14 USD
77156-068 77156-070 77156-072
1-Butyl-3-methylimidazolium methanesulfonate 98%
1-Butyl-3-methylimidazolium methanesulphonate

Caution: for Research Use Only

Formula: C₉H₁₈N₂O₃S
MDL Number: MFCD06798173
CAS Number: 342789-81-5
UN: 2923
ADR: 8,6.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR