Eosin Y 97% BS

Supplier: Ambeed
Warning

A308289-100G A308289-25G A308289-500G A308289-5G
77154-308EA 66 USD
77154-308 77154-310 77154-312 77154-314
Eosin Y 97% BS
Eosin Y

Caution: for Research Use Only

Formula: C₂₀H₈Br₄O₅
MDL Number: MFCD00036189
CAS Number: 15086-94-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR