H-Gln(isopropyl)-OH 95%

Supplier: Ambeed
Warning

A268722-100MG A268722-250MG
77153-600EA 15 USD
77153-600 77153-602
H-Gln(isopropyl)-OH 95%
H-Gln(isopropyl)-OH

Caution: for Research Use Only

Formula: C₈H₁₆N₂O₃
MDL Number: MFCD00672360
CAS Number: 4311-12-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR