(1S,4S)-2-Boc-2,5-diazabicyclo[2.2.1]heptane 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
tert-butyl (1S,4S)-2,5-diazabicyclo[2.2.1]heptane-2-carboxylate
A184819-1G
A184819-25G
A184819-100G
A184819-10G
A184819-250MG
A184819-5G
103815-788EA
9
USD
103815-788
103846-354
77083-976
77083-978
77151-730
77151-732
77151-734
77151-736
77301-920
(1S,4S)-2-Boc-2,5-diazabicyclo[2.2.1]heptane 98%
(1S,4S)-2-Boc-2,5-diazabicyclo[2.2.1]heptane
Caution: for Research Use Only
Formula:
C₁₀H₁₈N₂O₂ Melting Pt: 74…76 °C Flash Pt: 120.9 °C Storage Temperature: Ambient |
MDL Number:
MFCD01569250 CAS Number: 113451-59-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...