AZD1208 ≥98%

Supplier: Ambeed
Warning

Synonyms: (R,Z)-5-((2-(3-Aminopiperidin-1-yl)-[1,1'-biphenyl]-3-yl)methylene)thiazolidine-2,4-dione, AZD-1208

A178546-10MG A178546-1MG A178546-5MG
77151-556EA 30 USD
77151-556 77151-558 77151-560
AZD1208 ≥98%
AZD1208

Caution: for Research Use Only

Formula: C₂₁H₂₁N₃O₂S
MDL Number: MFCD25976757
CAS Number: 1204144-28-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR