Boc-D-Cha-OH 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Boc-β-cyclohexyl-D-Ala-OH
A126452-100G
A126452-250MG
A126452-1G
A126452-5G
103800-812EA
9
USD
103800-812
103831-668
77149-440
77149-442
77149-444
77298-124
77313-026
Boc-D-Cha-OH 98%
Boc-D-Cha-OH
Caution: for Research Use Only
Formula:
C₁₄H₂₅NO₄ MW: 271.357 g/mol Boiling Pt: 420.4 °C (760 mm Hg) Melting Pt: 64…67 °C Flash Pt: 208.1 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00133627 CAS Number: 127095-92-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...