MLN0128 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-(4-Amino-1-isopropyl-1H-pyrazolo[3,4-d]pyrimidin-3-yl)benzo[d]oxazol-2-amine, INK-128, Sapanisertib
A121801-10MG
A121801-2MG
A121801-5MG
A121801-1MG
A121801-100MG
A121801-50MG
A121801-250MG
77149-222EA
77
USD
77149-222
77149-224
77149-226
77611-036
77685-074
77685-075
77901-630
MLN0128 98%
MLN0128
Caution: for Research Use Only
Formula:
C₁₅H₁₅N₇O |
MDL Number:
MFCD22124893 CAS Number: 1224844-38-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...