Truxene 98%

Supplier: Ambeed
Warning

Synonyms: 10,15-Dihydro-5H-tribenzo[a,f,k]trindene, Tribenzylenebenzene, 10,15-Dihydro-5H-diindeno[1,2-a:1',2'-c]fluorene

A121220-100MG
77149-186EA 37.08 USD
77149-186
Truxene 98%
Truxene

Caution: for Research Use Only

Formula: C₂₇H₁₈
MW: 342.44 g/mol
MDL Number: MFCD00021180
CAS Number: 548-35-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR