AV-412 98+%
Supplier: Ambeed
|
Warning
|
A116496-10MG
A116496-1MG
A116496-25MG
A116496-2MG
A116496-5MG
A116496-50MG
77148-960EA
41
USD
77148-960
77148-962
77148-964
77148-966
77148-968
77685-031
AV-412 98+%
AV-412
Caution: for Research Use Only
Formula:
C₂₇H₂₈ClFN₆O |
MDL Number:
MFCD16038939 CAS Number: 451492-95-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...