VWR® Combination Refrigerator/Freezer
Supplier: VWR International
77148-140
77148-140EA 8677.1 USD
77148-140
VWR® Combination Refrigerator/Freezer
Refrigerators
These comprehensive cold storage units deliver next-level temperature control to laboratory environments.
  • Superior temperature uniformity
  • Microprocessor temperature controllers
  • Fully array of alarms
  • Forced draft air circulation
  • Natural hydrocarbon refrigerants
These combo units are designed with variable speed compressors to deliver unparalleled temperature control. Safeguard your samples with superior temperature uniformity and recovery, even after frequent door openings. Monitor your laboratory equipment though intuitive digital displays and protective alarms. Champion sustainability with natural hydrocarbon refrigerants - which not only contribute to energy savings, but also foster a greener laboratory footprint. Enjoy the convenience of auto-defrosting, eliminating the hassle of manual maintenance. *Elevate your laboratory cooling experience with an integrated solution that brings together innovation, performance and precision.
Certifications: ETL/cETL listed.
Caution: Auto-defrost freezers incorporate an electric heater installed on the evaporator that assists in the removal of the frost and ice. Although the heater helps speed up the frost and ice removal, it can raise the temperatures within the freezer. These air temperature spikes can be harmful to some medical and laboratory products.