CardioChek Plus Lipid Panel Sarter Kit With Printer
Supplier: PTS Diagnostics
PTS906
77148-112EA 1546.45 USD
77148-112
CardioChek Plus Lipid Panel Sarter Kit With Printer
Controls and Calibration Verification Materials
CardioChek Plus lipid panel starter kit with printer provides full lipid panel test results in as little as 90 seconds at the point of care.
Certifications: CLIA-waived, FDA-cleared, Meets NCEP guidelines for accuracy and precision, CRMLN-certified for CHOL and HDL tests.
Ordering information: CardioChek Plus lipid panel with printer starter kit includes: (1) CardioChek Plus analyzer, (4) PTS panels lipid panels (60 lipid tests), (1) PTS panels multi-chemistry controls, (4) PTS collect capillary tubes, 40 µl (64 capillary tubes total), (1) CardioChek printer (includes 1 roll of labels).