CardioChek Plus Lipid Panel Sarter Kit
Supplier: PTS Diagnostics
PTS905
77148-110EA 1271.85 USD
77148-110
CardioChek Plus Lipid Panel Sarter Kit
Controls and Calibration Verification Materials
CardioChek Plus lipid panel starter kit provides full lipid panel test results in as little as 90 seconds at the point of care.
Certifications: CLIA-waived, FDA-cleared, Meets NCEP guidelines for accuracy and precision, CRMLN-certified for CHOL and HDL tests.
Ordering information: CardioChek Plus lipid panel starter kit includes: (1) CardioChek Plus analyzer, (4) PTS panels lipid panels (60 lipid tests), (1) PTS panels multi-chemistry controls, (4) PTS collect capillary tubes, 40 µl (64 capillary tubes total).