CardioChek Plus Smart Bundle Starter Kit with Printer
Supplier: PTS Diagnostics
PTS761
77148-108EA 1664.16 USD
77148-108
CardioChek Plus Smart Bundle Starter Kit with Printer
Controls and Calibration Verification Materials
CardioChek Plus smart bundle starter kit with printer provides full lipid panel and glucose test results in as little as 90 seconds at the point of care.
Certifications: CLIA-waived, FDA-cleared, Meets NCEP guidelines for accuracy and precision, CRMLN-certified for CHOL and HDL tests.
Ordering information: CardioChek Plus smart bundle with printer starter kit includes: (1) CardioChek Plus analyzer, (4) PTS panels Lipid + eGlu smart bundles (60 lipid + glucose tests), (1) PTS panels multi-chemistry controls, (4) PTS collect capillary tubes, 40 µl (64 capillary tubes total), (1) CardioChek printer (includes 1 roll of labels).