CardioChek Plus Smart Bundle Starter Kit
Supplier: PTS Diagnostics
PTS760
77148-106EA 1445.29 USD
77148-106
CardioChek Plus Smart Bundle Starter Kit
Controls and Calibration Verification Materials
CardioChek Plus smart bundle starter kit provides full lipid panel and glucose test results in as little as 90 seconds at the point of care.
Certifications: CLIA-waived, FDA-cleared, Meets NCEP guidelines for accuracy and precision, CRMLN-certified for CHOL and HDL tests.
Ordering information: CardioChek Plus smart bundle starter kit includes: (1) CardioChek Plus analyzer, (4) PTS panels lipid + eGlu smart bundles (60 lipid + glucose tests), (1) PTS panels multi-chemistry controls, (4) PTS collect capillary tubes, 40 µl (64 capillary tubes total).