RightCare Disposable Bouffant Caps, 24", Spunbonded with Elastic Band

Supplier: OUTSOURCE PHARMACY SOLUTIONS, INC.
RCBFCPCA20
77148-080CS 122.4 USD
77148-080
RightCare Disposable Bouffant Caps, 24", Spunbonded with Elastic Band
Disposable Garments
RightCare 24" Bouffant caps provide basic protection against dirt, contaminants, and particulates in non hazardous environments.

  • White, non sterile bouffant caps
  • Highly breathable mesh fabric (10 to 12 g/m² spunbond polypropylene)
  • Caps provide generous airflow to keep the head cool
  • Dual elastic band is latex-free to reduce irritations

RightCare 24" Bouffant Caps are comfortable for workers to wear on a daily routine because they allows generous airflow to keep the head cool. Dual elastic band is latex-free and stretches easily to place the cap over the head for a snug fit. They are also effective at keeping hair contained to avoid contamination when used in various industries such as restaurants, food service, clean room environments, clinical settings, medical care facilities, tattoo product manufacturing, laboratories, hospitals, and pharmacies. Bouffant caps are also necessary to prevent cross-contamination for businesses that produce or package consumer products, pharmaceuticals, or assemble electronics.

Packaging: Caps are packaged 100 per bag.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR