α-Bromo-4-iodotoulene 98%
Supplier: Ambeed
|
Danger
|
A725429-10G
A725429-1G
A725429-25G
A725429-5G
77146-008EA
32.96
USD
77146-008
77146-010
77146-012
77146-014
α-Bromo-4-iodotoulene 98%
α-Bromo-4-iodotoulene
Caution: for Research Use Only
Formula:
C₇H₆BrI |
MDL Number:
MFCD00209656 CAS Number: 16004-15-2 UN: 3261 ADR: 8,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...