α-Bromo-4-iodotoulene 98%

Supplier: Ambeed
Danger

A725429-10G A725429-1G A725429-25G A725429-5G
77146-008EA 32.96 USD
77146-008 77146-010 77146-012 77146-014
α-Bromo-4-iodotoulene 98%
α-Bromo-4-iodotoulene

Caution: for Research Use Only

Formula: C₇H₆BrI
MDL Number: MFCD00209656
CAS Number: 16004-15-2
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR