(+)-(8,8-Dichlorocamphorylsulfonyl)oxaziridine 98%

Supplier: Ambeed
Danger

Synonyms: (4aS,7S,8aR)-8,8-Dichloro-9,9-dimethyltetrahydro-4H-4a,7-methanobenzo[c][1,2]oxazireno[2,3-b]isothiazole 3,3-dioxide

A604684-1G A604684-250MG A604684-25G A604684-5G
77144-894EA 33.99 USD
77144-894 77144-896 77144-898 77144-900
(+)-(8,8-Dichlorocamphorylsulfonyl)oxaziridine 98%
(+)-(8,8-Dichlorocamphorylsulfonyl)oxaziridine

Caution: for Research Use Only

Formula: C₁₀H₁₃Cl₂NO₃S
MDL Number: MFCD00243015
CAS Number: 127184-05-8
UN: 1479
ADR: 5.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR