(±)-3-Fluoro-α-methylbenzylamine 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-(3-Fluorophenyl)ethanamine
A586172-1G
A586172-250MG
77144-782EA
39.15
USD
77144-782
77144-784
(±)-3-Fluoro-α-methylbenzylamine 95%
(±)-3-Fluoro-α-methylbenzylamine
Caution: for Research Use Only
Formula:
C₈H₁₀FN MW: 139.17 g/mol |
MDL Number:
MFCD04038308 CAS Number: 74788-45-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...