L-Methionine 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-(-)-2-Amino-4-(methylthio)butyric acid, (S)-(-)-Methionine, L(-)-Methionine, (S)-(-)-2-Amino-4-(methylthio)butanoic acid, H-Met-OH, Methionine, (S)-2-Amino-4-(methylthio)butanoic acid
A351115-100G
A351115-25G
A351115-500G
A351115-1000G
77142-190EA
15.45
USD
77142-190
77142-192
77142-194
77685-996
L-Methionine 95%
L-Methionine
Caution: for Research Use Only
Formula:
C₅H₁₁NO₂S MW: 149.21 g/mol Boiling Pt: 278 Melting Pt: 136 |
MDL Number:
MFCD00063097 CAS Number: 63-68-3 Merck Index: 13,06004 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...