L-Methionine 95%

Supplier: Ambeed
Warning

Synonyms: (S)-(-)-2-Amino-4-(methylthio)butyric acid, (S)-(-)-Methionine, L(-)-Methionine, (S)-(-)-2-Amino-4-(methylthio)butanoic acid, H-Met-OH, Methionine, (S)-2-Amino-4-(methylthio)butanoic acid

A351115-100G A351115-25G A351115-500G A351115-1000G
77142-190EA 15.45 USD
77142-190 77142-192 77142-194 77685-996
L-Methionine 95%
L-Methionine

Caution: for Research Use Only

Formula: C₅H₁₁NO₂S
MW: 149.21 g/mol
Boiling Pt: 278
Melting Pt: 136
MDL Number: MFCD00063097
CAS Number: 63-68-3
Merck Index: 13,06004

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR