3,4-Difluorobenzoic acid 97%

Supplier: Ambeed
Warning

A306986-1000G A306986-25G A306986-100G A306986-10G A306986-500G
103810-046EA 206 USD
103810-046 76838-398 76838-400 77141-624 77141-626 77141-628
3,4-Difluorobenzoic acid 97%

Caution: for Research Use Only

Formula: C₇H₄F₂O₂
Boiling Pt: 257 °C (760 mm Hg)
Melting Pt: 120…125 °C
Flash Pt: 109.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00011672
CAS Number: 455-86-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR