2-Acetamido-6-hydroxypurine 98%
Supplier: Ambeed
|
Warning
|
A285763-10G
A285763-1G
A285763-5G
A285763-100G
77141-244EA
60.78
USD
77141-244
77141-246
77141-248
77550-756
2-Acetamido-6-hydroxypurine 98%
2-Acetamido-6-hydroxypurine
Caution: for Research Use Only
Formula:
C₇H₇N₅O₂ |
MDL Number:
MFCD00078201 CAS Number: 19962-37-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...