4-Bromo-m-cresol 98%

Supplier: Ambeed
Warning

A164627-100G A164627-10G A164627-500G A164627-25G A164627-5G
103844-142EA 6 USD
103844-142 77138-274 77138-276 77138-278 77300-728 77320-326
4-Bromo-m-cresol 98%
4-Bromo-m-cresol

Caution: for Research Use Only

Formula: C₇H₇BrO
Boiling Pt: 142…145 °C (760 mm Hg)
Melting Pt: 59…61 °C
Storage Temperature: Ambient
MDL Number: MFCD00079723
CAS Number: 14472-14-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR