4-Nitro-2-(trifluoromethyl)aniline 98%

Supplier: Ambeed
Warning

A159502-100G A159502-10G A159502-25G A159502-5G
77138-070EA 33.99 USD
77138-070 77138-072 77138-074 77138-076 77138-078
4-Nitro-2-(trifluoromethyl)aniline 98%
4-Nitro-2-(trifluoromethyl)aniline

Caution: for Research Use Only

Formula: C₇H₅F₃N₂O₂
MDL Number: MFCD00007365
CAS Number: 121-01-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR