5-Amino-2-chlorobenzoic acid 98%

Supplier: Ambeed
Warning

A159259-100G A159259-10G A159259-25G A159259-5G
77138-062EA 51.51 USD
77138-062 77138-064 77138-066 77138-068
5-Amino-2-chlorobenzoic acid 98%
5-Amino-2-chlorobenzoic acid

Caution: for Research Use Only

Formula: C₇H₆ClNO₂
MDL Number: MFCD00007845
CAS Number: 89-54-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR