3-Fluoro-4-nitrobenzoic acid 98%

Supplier: Ambeed
Warning

A123794-100G A123794-10G A123794-25G A123794-500G A123794-1G A123794-5G
103800-444EA 9 USD
103800-444 103831-282 77136-614 77136-616 77136-618 77136-620 77297-958 77312-564
3-Fluoro-4-nitrobenzoic acid 98%
3-Fluoro-4-nitrobenzoic acid

Caution: for Research Use Only

Formula: C₇H₄FNO₄
Boiling Pt: 372.8 °C (760 mm Hg)
Melting Pt: 174…175 °C
Flash Pt: 179.3 °C
Storage Temperature: Ambient
MDL Number: MFCD01862092
CAS Number: 403-21-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR