3-Fluoro-4-nitrobenzoic acid 98%
Supplier: Ambeed
|
Warning
|
A123794-100G
A123794-10G
A123794-25G
A123794-500G
A123794-1G
A123794-5G
103800-444EA
9
USD
103800-444
103831-282
77136-614
77136-616
77136-618
77136-620
77297-958
77312-564
3-Fluoro-4-nitrobenzoic acid 98%
3-Fluoro-4-nitrobenzoic acid
Caution: for Research Use Only
Formula:
C₇H₄FNO₄ Boiling Pt: 372.8 °C (760 mm Hg) Melting Pt: 174…175 °C Flash Pt: 179.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD01862092 CAS Number: 403-21-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...