4-Carboxyphenylboronic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Boronobenzoic Acid, Ark Pharm, 4-Carboxybenzeneboronic acid
A109648-100G
A109648-10G
A109648-1G
A109648-25G
A109648-500G
A109648-5G
103823-340EA
6
USD
103823-340
77135-920
77135-922
77135-924
77135-926
77135-928
77310-150
4-Carboxyphenylboronic acid 98%
4-Carboxyphenylboronic acid
Caution: for Research Use Only
Formula:
C₇H₇BO₄ Boiling Pt: 406 °C (760 mm Hg) Melting Pt: 220 °C Flash Pt: 150.4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00151801 CAS Number: 14047-29-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...