Digital Homogenizers, OV 625
Supplier: VELP SCIENTIFIC INC.
SA20910470 F20910470
77119-176EA 4370.9 USD
77119-176 77119-174
Digital Homogenizers, OV 625
Homogenizers Benchtop Homogenizers
Powerful digital homogenizer designed for the maximum level of reproducibility in a wide range of applications. Cutting-edge technology and engineering expertise guarantee precision and high shearing force for fast dispersion, homogenization, emulsification, suspensions, and high-speed mixing of biological tissues, pharmaceuticals, cosmetics and food products.
  • Volume range from 0.2 ml to 2.5 L (H₂O), handles samples up to 5000 mPas viscosity
  • Bright 3-digit LED display and the comfortable rotating knob enable precise electronic speed settings from 3000 to 25000 min
  • Various safety functions, such as a soft start, motor overload protection and overtemperature alarms ensure the highest safety in daily operations
  • Resistant and robust housing protects the internal electronic parts and ensures the instrument works even in the most challenging applications
  • Premium construction materials guarantee excellent mechanical properties and chemical resistance
  • Wide range of dispersing tools ensures excellent performance for many applications for small samples and large volumes
  • Fast and easy installation system enables to easily accommodate and change the tools
The OV 625 Digital incorporates cutting-edge technologies and high levels of engineering expertise, to guarantee high shearing force and excellent homogenizing, emulsifying, suspending and mixing performance for a wide range of applications. Thanks to a wide range of dispersing tools, the OV 625 Digital enables processing volumes ranging from 0.2 ml to 2.5 L (H₂O) and up to 5000 mPas viscous samples.
The control panel is designed to optimize the user experience: The speed parameter is visible at a glance to provide excellent visibility in every working condition. Various safety features provide maximum protection for the user in daily operations. Once the set point speed is set, the OV 625 Digital ensures a soft start-up to reach the desired speed gradually. In addition, several alarms inform the operator in case of overload and overtemperature.
The resistant and rugged housing protects all internal electronic components and enables proper operations in the most challenging conditions. The ergonomic, compact and innovative design and the robust structure make the OV 625 Digital a solution designed to last, guaranteed by a three-year warranty (online product registration required).
Choosing the proper dispersing tool is crucial to achieve the best performance and high reproducibility. A wide range of dispersing tools ensures excellent performance for many applications for small samples, sizes such as biological matrices, and large volumes (up to 4 L as a high-speed mixer).
The VELP dispersing tools are manufactured with premium construction materials to offer great compatibility, chemical resistance to various samples and reagents, and optimal mechanical properties. Moreover, a fast and easy installation system enables you to easily accommodate and change the tools, getting ready for the next application is a breeze.
Certifications: CB; cNEMKOus.