Scopoletin (7-hydroxy-6-methoxycumarin) 97%

Supplier: Ambeed
Warning

A867040-100MG A867040-10MG A867040-1G A867040-1MG A867040-250MG A867040-50MG
77118-626EA 51.51 USD
77118-626 77118-628 77118-630 77118-632 77118-634 77118-636
Scopoletin (7-hydroxy-6-methoxycumarin) 97%
Scopoletin (7-Hydroxy-6-methoxycumarin)

Caution: for Research Use Only

Formula: C₁₀H₈O₄
MDL Number: MFCD00006872
CAS Number: 92-61-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR