5-Acetylvaleric acid 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
6-Oxoheptanoic acid
A597192-10G
A597192-1G
A597192-250MG
A597192-25G
A597192-5G
77117-240EA
62.83
USD
77117-240
77117-242
77117-244
77117-246
77117-248
5-Acetylvaleric acid 98%
5-Acetylvaleric acid
Caution: for Research Use Only
Formula:
C₇H₁₂O₃ |
MDL Number:
MFCD00051479 CAS Number: 3128-07-2 UN: 3261 ADR: 8,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...