9,10-Bis(phenylethynyl)anthracene 95%

Supplier: Ambeed
Warning

A541261-100G A541261-10G
77116-826EA 199.81 USD
77116-826 77116-828
9,10-Bis(phenylethynyl)anthracene 95%
9,10-Bis(phenylethynyl)anthracene

Caution: for Research Use Only

Formula: C₃₀H₁₈
MW: 378.47 g/mol
MDL Number: MFCD00012050
CAS Number: 10075-85-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR